missing translation for 'onlineSavingsMsg'
Learn More

BET-BAY 002 (S enantiomer), MedChemExpress

Product Code. 16447877
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447877 5 mg 5mg
16490156 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16447877 Supplier Medchem Express Supplier No. HY12421B

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress BET-BAY 002 S enantiomer is the S-enantiomer of BET-BAY 002. BET-BAY 002 is a BET inhibitor.

BET-BAY 002 S enantiomer is the S-enantiomer of BET-BAY 002. BET-BAY 002 is a BET inhibitor.

Specifications

Chemical Name or Material BET-BAY 002 (S enantiomer)
Physical Form Solid
CAS 2070009-49-1
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H18ClN5O
For Use With (Application) Neuroscience-Neuromodulation
SMILES CC1=NN=C2N1C3=CC=CC=C3C(C4=CC=C(Cl)C=C4)=C[C@@H]2CC5=NN=C(C)O5
Molecular Weight (g/mol) 403.86
Formula Weight 403.86
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.