missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-Alpha,Alpha-d2 Alcohol, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Benzyl-Alpha,Alpha-d2 Alcohol |
| CAS | 21175-64-4 |
| Molecular Formula | C7H6D2O |
| Quantity | 5 mg |
| Synonym | Benzyl-α,α-d2 alcohol (6CI','7CI','8CI), 1,1-d2-Phenylmethanol, [α,α-2H2]Benzyl alcohol, Benzenemethanol-α,α-d2, α,α-Dideuteriobenzyl alcohol |
| InChI Formula | InChI=1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2/i6D2 |
| SMILES | OC([2H])([2H])C1=CC=CC=C1 |
| IUPAC Name | phenylmethan-d2-ol |
| Molecular Weight (g/mol) | 110.15 |
| Formula Weight | 110.07 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction