missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,7,8-Tetrachloro-p-dioxin, TRC
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | 2,3,7,8-Tetrachlorodibenzo-p-dioxin |
| CAS | 1746-01-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 H4 Cl4 O2 |
| Quantity | 5 mg |
| Synonym | Dibenzo[b,e][1,4]dioxin','2,3,7,8-tetrachloro-, Dibenzo-p-dioxin','2,3,7,8-tetrachloro- (6CI,7CI,8CI), 2,3,7,8-Tetrachlorodibenzo[b,e][1,4]dioxin, 2,3,7,8-TCDD, 2,3,7,8-Tetrachloro-p-dioxin, 2,3,7,8-Tetrachlorodibenzo-1,4-dioxin, 2,3,7,8-Tetrachlorodibenzo-p-dioxin, 2,3,7,8-Tetrachlorodibenzodioxin, 2,3,7,8-tetrachlorodibenzo-p-dioxin, 2378TCDD, D 48, Dioxin, Dioxin (herbicide contaminant), PCDD 48, TCDBD, TCDD, Tetrachlorodibenzo-p-dioxin, Tetrachlorodibenzodioxin |
| InChI Formula | InChI=1S/C12H4Cl4O2/c13-5-1-9-10(2-6(5)14)18-12-4-8(16)7(15)3-11(12)17-9/h1-4H |
| SMILES | Clc1cc2Oc3cc(Cl)c(Cl)cc3Oc2cc1Cl |
| IUPAC Name | 2,3,7,8-tetrachlorodibenzo-p-dioxin |
| Molecular Weight (g/mol) | 321.97 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction