missing translation for 'onlineSavingsMsg'
Learn More

17Alpha-Vinyltestosterone Acryl Ester, TRC

Product Code. 30449596
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 30449596

Brand: Toronto Research Chemicals TRCV5200035MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17alpha-Vinyltestosterone Acryl Ester
Analyte or Component Names 17alpha-Vinyltestosterone Acryl Ester
Shipping Condition Room Temperature
Molecular Formula C24H32O3
Synonym (8R,9S,10R,13S,14S,17R)-10,13-Dimethyl-3-oxo-17-vinyl-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl Acrylate
InChI Formula InChI=1S/C24H32O3/c1-5-21(26)27-24(6-2)14-11-20-18-8-7-16-15-17(25)9-12-22(16,3)19(18)10-13-23(20,24)4/h5-6,15,18-20H,1-2,7-14H2,3-4H3/t18-,19+,20+,22+,23+,24+/m1/s1
Solution Type Neat
SMILES O=C1CC[C@@]2(C)C(CC[C@]3([H])[C@]2([H])CC[C@](C)([C@@](C=C)(OC(C=C)=O)CC4)[C@]34[H])=C1
IUPAC Name (8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3-oxo-17-vinyl-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl acrylate
Industry Type Forensics
Molecular Weight (g/mol) 368.51
Quantity 5 mg
Formula Weight 368.24
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.