missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium-d6 Inner Salt, TRC
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Specifications
Specifications
| Chemical Name or Material | 1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium-d6 Inner Salt |
| Molecular Formula | C19 D6 H17 N O3 S |
| Quantity | 5 mg |
| Synonym | 1,1,2-Trimethyl-3-(4-sulfobutyl)-1H-benz[e]indolium Inner Salt-d6, 1,1,2-Trimethyl-3-(4-sulfobutyl)-1H-benz[e]indolium Hydroxide Inner Salt-d6, 4-(1,1,2-Trimethyl-1H-benzo[e]indolium-3-yl)butane-1-sulfonate-d6 |
| InChI Formula | InChI=1S/C19H23NO3S/c1-14-19(2,3)18-16-9-5-4-8-15(16)10-11-17(18)20(14)12-6-7-13-24(21,22)23/h4-5,8-11H,6-7,12-13H2,1-3H3/i2D3,3D3 |
| SMILES | [2H]C([2H])([2H])C1(C(=[N+](CCCCS(=O)(=O)[O-])c2ccc3ccccc3c12)C)C([2H])([2H])[2H] |
| IUPAC Name | 4-[2-methyl-1,1-bis(trideuteriomethyl)benzo[e]indol-3-ium-3-yl]butane-1-sulfonate |
| Molecular Weight (g/mol) | 351.493 |
| Formula Weight | 351.178 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction