missing translation for 'onlineSavingsMsg'
Learn More

HUBER 5L Thermofluid

Product Code. 10775016
Change view
Click to view available options
Temperature Range:
-10°C to +120°C
-20°C to 195°C to 235°C
-40°C to 165°C to 220°C
-60°C to 115°C to 200°C
-80°C to +55°C
-80°C to 100°C to 250°C
-90°C to +55°C to 170°C
20°C to 275°C
Temperature (Metric) Maximum:
100°C, 250°C
115°C, 200°C
120°C
165°C, 220°C
195°C, 235°C
275°C
55°C
55°C, 170°C
Viscosity:
10 mm2/sec.
20 mm2/sec.
3 mm2/sec.
5 mm2/sec.
50 mm2/sec.
8 mm2/sec.
Type:
Silicon Oil
Synthetic Oil
Unit Size:
5 L
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range Temperature (Metric) Maximum Viscosity Type unitSize
10775016 -10°C to +120°C 120°C 8 mm2/sec. Synthetic Oil 5 L
11517364 -20°C to 195°C to 235°C 195°C, 235°C 20 mm2/sec. Silicon Oil 5 L
11527424 -40°C to 165°C to 220°C 165°C, 220°C 10 mm2/sec. Silicon Oil 5 L
11587364 -80°C to 100°C to 250°C 100°C, 250°C 3 mm2/sec. Silicon Oil 5 L
11547364 -60°C to 115°C to 200°C 115°C, 200°C 5 mm2/sec. Silicon Oil 5 L
11567364 -80°C to +55°C 55°C 3 mm2/sec. Silicon Oil 5 L
11517374 -90°C to +55°C to 170°C 55°C, 170°C 3 mm2/sec. Silicon Oil 5 L
11547424 20°C to 275°C 275°C 50 mm2/sec. Silicon Oil 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10775016 Supplier HUBER Supplier No. 9684
  • ONLINE EXCLUSIVE PRICE
    £514.00
    £395.00 / 5 L
    Save £119.00
    23% Off
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Recommended for many temperature control applications. Huber Kaeltmaschinenbau™ 5L Thermofluid oils provide optimal heat transfer. Temperature ranges vary from -90° – 275°C, with viscosity at 25° ranging from 3 to 50mm2/second.

  • Thermofluids for many temperature control applications
  • Display the best possible thermodynamic and environmental characteristics
  • Safe reliable operation relies on compliance with safety standards to ensure optimal results
  • Many varieties have higher maximum temperature when used in a Unistat™ closed application
  • Convenient 5L cans of Silicon or Synthetic Oil for limited storage or work spaces

Specifications

Temperature Range -10°C to +120°C
Type Synthetic Oil
Quantity 5 L
Temperature (Metric) Maximum 120°C
Viscosity 8 mm2/sec.
compliance-icons compliance-icons
Product Identifier
  • synoil M10.120.08
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
Hazard Statement
  • H332-Harmful if inhaled.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1-decene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.