Learn More
Smeg 5L Liquid alkaline detergent DETERLIQUID I2
Brand: Smeg DETERLIQUIDI2
5L Liquid alkaline detergent DETERLIQUID I2
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P310-Immediately call a POISON CENTER/doctor/.
- P351-Rinse cautiously with water for several minutes.
Supplemental information
- MIXTURE LIST-Contain: potasssium iodide
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.