missing translation for 'onlineSavingsMsg'
Learn More

5H-Dibenzo[a,d]cycloheptene, TRC

Product Code. 30138082
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138082 2.5 g 2.5g
30009662 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138082 Supplier Toronto Research Chemicals Supplier No. TRCD4170552.5G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256-81-5
Molecular Formula C15 H12
Molecular Weight (g/mol) 192.26
Synonym 5H-Dibenzo[a,d][7]annulene, Cyproheptadine Hydrochloride Imp. A (EP), Dibenzocycloheptene
IUPAC Name 11H-dibenzo[1,2-b:2',1'-e][7]annulene
SMILES C1c2ccccc2C=Cc3ccccc13

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C15H12/c1-3-7-14-11-15-8-4-2-6-13(15)10-9-12(14)5-1/h1-10H,11H2
Formula Weight 192.0939
Chemical Name or Material Dibenzocycloheptene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.