missing translation for 'onlineSavingsMsg'
Learn More

5H,6H,7H,8H-Imidazo[1,2-a]pyridine-3-sulfonyl Chloride (~85%), TRC

Product Code. 30397529
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397529 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397529 Supplier Toronto Research Chemicals Supplier No. TRCH32568350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5H,6H,7H,8H-Imidazo[1,2-a]pyridine-3-sulfonyl Chloride
CAS 1216892-47-5
Molecular Formula C7H9N2O2SCl
Quantity 50 mg
Synonym 5,6,7,8-Tetrahydroimidazo[1,2-a]-pyridine-3-sulfonyl chloride
InChI Formula InChI=1S/C7H9ClN2O2S/c8-13(11,12)7-5-9-6-3-1-2-4-10(6)7/h5H,1-4H2
SMILES ClS(=O)(=O)c1cnc2CCCCn12
IUPAC Name 5,6,7,8-tetrahydroimidazo[1,2-a]pyridine-3-sulfonyl chloride
Molecular Weight (g/mol) 220.67
Formula Weight 220.0073
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.