missing translation for 'onlineSavingsMsg'
Learn More

Vinylphosphonic Acid, TRC

Product Code. 30451571
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30451571

Brand: Toronto Research Chemicals TRCV4534155G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vinylphosphonic Acid
CAS 1746-03-8
Molecular Formula C2H5O3P
Quantity 5 g
Synonym P-Ethenylphosphonic Acid, Ethenephosphonic Acid, Ethylenephosphonic Acid ITC 467 Vinylphosphonic Acid, Ethenyl-Phosphonic Acid
InChI Formula InChI=1S/C2H5O3P/c1-2-6(3,4)5/h2H,1H2,(H2,3,4,5)
SMILES OP(C=C)(O)=O
IUPAC Name ethenylphosphonic acid
Molecular Weight (g/mol) 108.03
Formula Weight 107.9976
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.