missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium(IV) Oxide Acetylacetonate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Vanadium(IV) Oxide Acetylacetonate |
| CAS | 3153-26-2 |
| Molecular Formula | C10H14O5V |
| Quantity | 5 g |
| Synonym | Bis(2,4-pentanedionato)oxovanadium, Bis(2,4-pentanedionato)oxovanadium(IV), Bis(acetylacetonato)oxovanadium, Bis(acetylacetonato)oxovanadium(IV), Bis(acetylacetonato)oxyvanadium, NSC 116105, NSC 4659, NSC 52327, Nacem Vanadyl, Oxobis(2,4-pentanedionato)vanadium, Oxobis(acetylacetonato)vanadium, Oxovanadium(II) Acetylacetonate, Vanadium Oxide Acetylacetonate, Vanadium(IV) Bis(acetylacetonato)(oxo), Vanadium(IV) Oxide Bis(2,4-pentanedionate), Vanadyl Acetylacetonate, Vanadyl Bis(2,4-pentanedionate), Vanadyl Bis(acetylacetonate), VoAA |
| InChI Formula | InChI=1S/2C5H8O2.O.V/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;;/q;;;+2/p-2/b2*4-3-;; |
| SMILES | CC(=O)\C=C(\C)/O[V](=O)O\C(=C/C(=O)C)\C |
| IUPAC Name | oxo-bis[[(Z)-4-oxopent-2-en-2-yl]oxy]vanadium |
| Molecular Weight (g/mol) | 265.16 |
| Formula Weight | 265.0281 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction