missing translation for 'onlineSavingsMsg'
Learn More

(5EZ,10RS)-5-[3-(Dimethyl-amino)propylidene]-10,11-dihydro-5H-dibenzo[a,d][7]annulen-10-ol, Mikromol™

Product Code. 17973236
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17973236 25 mg 25mg
17921066 100 mg 100mg
30459938 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973236 Supplier Mikromol Supplier No. MM0062.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(5EZ,10RS)-5-[3-(Dimethyl-amino)propylidene]-10,11-dihydro-5H-dibenzo[a,d][7]annulen-10-ol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10-Hydroxyamitriptyline
CAS 1159-82-6
Analyte or Component Names 10-Hydroxyamitriptyline
Shipping Condition Room Temperature
Molecular Formula C20 H23 N O
Certifications/Compliance ISO 9001
Synonym Amitriptyline Hydrochloride Imp. F (EP), Amitriptyline Imp. F (EP), (5EZ,10RS)-5-[3-(Dimethyl-amino)propylidene]-10,11-dihydro-5H-dibenzo[a,d][7]annulen-10-ol, 10-Hydroxyamitriptyline, Amitriptyline Hydrochloride Impurity F, Amitriptyline Impurity F
InChI Formula InChI=1S/C20H23NO/c1-21(2)13-7-12-17-16-9-4-3-8-15(16)14-20(22)19-11-6-5-10-18(17)19/h3-6,8-12,20,22H,7,13-14H2,1-2H3
SMILES CN(C)CCC=C1c2ccccc2CC(O)c3ccccc13
IUPAC Name 11-[3-(dimethylamino)propylidene]-5,6-dihydrodibenzo[1,2-a:2',1'-d][7]annulen-5-ol
Molecular Weight (g/mol) 293.4
Quantity 25 mg
Formula Weight 293.178 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.