missing translation for 'onlineSavingsMsg'
Learn More

5Beta-Dihydrocortisone Acetate 21-Acetate, TRC

Product Code. 30383289
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383289 10 mg
30375301 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383289 Supplier Toronto Research Chemicals Supplier No. TRCD44884010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1499-59-8
Molecular Formula C23H32O6
Molecular Weight (g/mol) 404.5
Synonym 17,21-Dihydroxy-5beta-pregnane-3,11,20-trione 21-Acetate, L 491297-0, NSC 22843
IUPAC Name [2-[(5R,8S,9S,10S,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-1,2,4,5,6,7,8,9,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CC[C@@H]4CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H32O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h14,16-17,20,28H,4-12H2,1-3H3/t14-,16+,17+,20-,21+,22+,23+/m1/s1
Formula Weight 404.2199
Chemical Name or Material 5beta-Dihydrocortisone Acetate 21-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.