missing translation for 'onlineSavingsMsg'
Learn More

(5Beta,7Beta)-7-Hydroxy-3-oxo-24-norcholan-23-oic Acid Methyl Ester, TRC

Product Code. 30402623
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402623 10 mg 10mg
30397427 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402623 Supplier Toronto Research Chemicals Supplier No. TRCH94901510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5beta,7beta)-7-Hydroxy-3-oxo-24-norcholan-23-oic Acid Methyl Ester
Analyte or Component Names (5beta,7beta)-7-Hydroxy-3-oxo-24-norcholan-23-oic Acid Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C24H38O4
InChI Formula InChI=1S/C24H38O4/c1-14(11-21(27)28-4)17-5-6-18-22-19(8-10-24(17,18)3)23(2)9-7-16(25)12-15(23)13-20(22)26/h14-15,17-20,22,26H,5-13H2,1-4H3/t14-,15+,17-,18+,19+,20+,22+,23+,24-/m1/s1
Solution Type Neat
SMILES [H][C@@]12[C@@H](O)C[C@]3([H])CC(CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4([H])[C@H](C)CC(OC)=O)=O
IUPAC Name methyl (R)-3-((5R,7S,8R,9S,10S,13R,14S,17R)-7-hydroxy-10,13-dimethyl-3-oxohexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)butanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 390.56
Quantity 10 mg
Formula Weight 390.28
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.