missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5Beta/6Beta-Hydroxy Lurasidone Hydrochloride(Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | 5beta/6beta-Hydroxy Lurasidone Hydrochloride(Mixture of Diastereomers) |
| Analyte or Component Names | 5beta/6beta-Hydroxy Lurasidone Hydrochloride(Mixture of Diastereomers) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28H37ClN4O3S |
| Synonym | [2(1R*,2R*),3aα,4β,(5/6)β,7β,7aα]-2-[[2-[[4-(1,2-Benzisothiazol-3-yl)-1-piperazinyl]methyl]cyclohexyl]methyl]hexahydro-(5/6)-hydroxy-4,7-methano-1H-isoindole-1,3(2H)-dione Hydrochloride, Lurasidone Metabolite M-8 Hydrochloride |
| InChI Formula | InChI=1S/2C28H36N4O3S.2ClH/c2*33-22-14-19-13-21(22)25-24(19)27(34)32(28(25)35)16-18-6-2-1-5-17(18)15-30-9-11-31(12-10-30)26-20-7-3-4-8-23(20)36-29-26;;/h2*3-4,7-8,17-19,21-22,24-25,33H,1-2,5-6,9-16H2;2*1H/t17-,18-,19+,21-,22-,24+,25-;17-,18-,19-,21+,22+,24-,25+;;/m00../s1 |
| Solution Type | Neat |
| SMILES | [H][C@]([C@@]1([H])C2=O)([C@@]3([H])[C@H](O)C[C@]1([H])C3)C(N2C[C@@H]4CCCC[C@H]4CN5CCN(C6=NSC7=C6C=CC=C7)CC5)=O.[H][C@]([C@@]8([H])C9=O)([C@@]%10([H])C[C@H](O)[C@]8([H])C%10)C(N9C[C@@H]%11CCCC[C@H]%11CN%12CCN(C%13=NSC%14=C%13C=CC=C%14)CC%12)=O.Cl.Cl |
| IUPAC Name | (3aR,4S,5R,7S,7aS)-2-(((1R,2R)-2-((4-(benzo[d]isothiazol-3-yl)piperazin-1-yl)methyl)cyclohexyl)methyl)-5-hydroxyhexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione--(3aS,4R,5S,7R,7aR)-2-(((1R,2R)-2-((4-(benzo[d]isothiazol-3-yl)piperazin-1-yl)methyl)cyclohexyl)methyl)-5-hydroxyhexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione (1/1) dihydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?