missing translation for 'onlineSavingsMsg'
Learn More

(5Beta,6Alpha)-6-Hydroxy-3-oxo-cholan-24-oic Acid, TRC

Product Code. 30389533
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389533 5 mg 5mg
30400601 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389533 Supplier Toronto Research Chemicals Supplier No. TRCH9490305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67733-54-4
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.56
Synonym 6alpha-Hydroxy-3-oxo-5beta-cholan-24-oic Acid
IUPAC Name (4R)-4-[(5R,6S,8S,9S,10R,13R,14S,17R)-6-hydroxy-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3C[C@H](O)[C@@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C24H38O4/c1-14(4-7-22(27)28)17-5-6-18-16-13-21(26)20-12-15(25)8-10-24(20,3)19(16)9-11-23(17,18)2/h14,16-21,26H,4-13H2,1-3H3,(H,27,28)/t14-,16+,17-,18+,19+,20+,21+,23-,24-/m1/s1
Formula Weight 390.277
Chemical Name or Material (5beta,6alpha)-6-Hydroxy-3-oxo-cholan-24-oic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.