missing translation for 'onlineSavingsMsg'
Learn More

5b-Dihydro Progesterone, TRC

Product Code. 30385682
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385682 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385682 Supplier Toronto Research Chemicals Supplier No. TRCD44928050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5beta-Dihydro Progesterone
CAS 128-23-4
Molecular Formula C21 H32 O2
Quantity 50 mg
Synonym Pregnane-3,20-dione, (5β)-, 5β-Pregnane-3,20-dione (8CI), (5β)-Pregnane-3,20-dione, 5β-Dihydro progesterone, 5β-Dihydroprogesterone, 5β-Pregnan-3,20-dione, NSC 82868, U 2411
InChI Formula InChI=1S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h14,16-19H,4-12H2,1-3H3/t14-,16+,17-,18+,19+,20+,21-/m1/s1
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (5R,8R,9S,10S,13S,14S,17S)-17-acetyl-10,13-dimethyl-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 316.48
Formula Weight 316.2402
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.