missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5aR,6R)-6-[[(Z)-(Furan-2-yl)(methoxyimino)acetyl]amino]-5a,6-dihydro-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazine-1,7(4H)-dione, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(5aR,6R)-6-[[(Z)-(Furan-2-yl)(methoxyimino)acetyl]amino]-5a,6-dihydro-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazine-1,7(4H)-dione
Specifications
Specifications
| Chemical Name or Material | (alphaZ)-alpha-(Methoxyimino)-N-[(5aR,6R)-1,4,5a,6-tetrahydro-1,7-dioxo-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazin-6-yl]-2-furanacetamide |
| CAS | 947723-87-7 |
| Analyte or Component Names | (alphaZ)-alpha-(Methoxyimino)-N-[(5aR,6R)-1,4,5a,6-tetrahydro-1,7-dioxo-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazin-6-yl]-2-furanacetamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H13 N3 O6 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | (αZ)-α-(Methoxyimino)-N-[(5aR,6R)-1,4,5a,6-tetrahydro-1,7-dioxo-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazin-6-yl]-2-furanacetamide, Cefuroxime Imp. H (EP), Cefuroxime Axetil Imp. E (EP), Cefuroxime Axetil Impurity E, Cefuroxime Sodium Impurity H, Cefuroxime Impurity H |
| InChI Formula | InChI=1S/C15H13N3O6S/c1-22-17-9(8-3-2-4-23-8)12(19)16-10-13(20)18-11-7(5-24-15(11)21)6-25-14(10)18/h2-4,10,14H,5-6H2,1H3,(H,16,19)/b17-9-/t10-,14-/m1/s1 |
| SMILES | CO\N=C(/C(=O)N[C@H]1[C@H]2SCC3=C(N2C1=O)C(=O)OC3)\c4occc4 |
| Molecular Weight (g/mol) | 363.35 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction