missing translation for 'onlineSavingsMsg'
Learn More

5Alpha-Dihydrolevonorgestrel, TRC

Product Code. 30385280
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385280 10 mg
30374548 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385280 Supplier Toronto Research Chemicals Supplier No. TRCD44966010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78088-19-4
Molecular Formula C21 H30 O2
Molecular Weight (g/mol) 314.46
Synonym 18,19-Dinorpregn-20-yn-3-one, 13-ethyl-17-hydroxy-, (5α,17α)- (9CI), 5α-Dihydrolevonorgestrel, (5α,17α)-13-Ethyl-17-hydroxy-18,19-dinorpregn-20-yn-3-one, 13-Ethyl-17-hydroxy-18,19-dinor-5α,17α-pregn-20-yn-3-one
IUPAC Name (5S,8R,9R,10S,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-1,2,4,5,6,7,8,9,10,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-3-one
SMILES CC[C@]12CC[C@@H]3[C@H]4CCC(=O)C[C@@H]4CC[C@H]3[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H30O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,14,16-19,23H,3,5-13H2,1H3/t14-,16-,17+,18+,19-,20-,21-/m0/s1
Formula Weight 314.2246
Chemical Name or Material 5alpha-Dihydrolevonorgestrel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.