missing translation for 'onlineSavingsMsg'
Learn More

5Alpha-Dihydrocortisol 21-Tetrahydro-2H-pyran-2-yl, TRC

Product Code. 30381794
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381794 500 mg 500mg
30372121 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381794 Supplier Toronto Research Chemicals Supplier No. TRCD450395500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5alpha-Dihydrocortisol 21-Tetrahydro-2H-pyran-2-yl
CAS 131061-49-9
Molecular Formula C26H40O6
Quantity 500 mg
Synonym (5α,11β)-11,17-dihydroxy-21-[(tetrahydro-2H-pyran-2-yl)oxy]-pregnane-3,20-dione
InChI Formula InChI=1S/C26H40O6/c1-24-10-8-17(27)13-16(24)6-7-18-19-9-11-26(30,25(19,2)14-20(28)23(18)24)21(29)15-32-22-5-3-4-12-31-22/h16,18-20,22-23,28,30H,3-15H2,1-2H3/t16-,18-,19-,20-,22?,23+,24-,25-,26-/m0/s1
SMILES C[C@@]12[C@]([C@]3([C@]([C@@H](O)C1)([C@]4(C)[C@@](CC3)(CC(=O)CC4)[H])[H])[H])(CC[C@@]2(C(COC5CCCCO5)=O)O)[H]
IUPAC Name (5S,8S,9S,10S,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-17-(2-((tetrahydro-2H-pyran-2-yl)oxy)acetyl)hexadecahydro-3H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 448.59
Formula Weight 448.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.