missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5Alpha-Dihydro Testosterone Undecanoate (1.0mg/ml in Acetonitrile), TRC
Specifications
Specifications
| CAS | 6804-12-2 |
| Molecular Formula | C30H50O3 |
| Quantity | 10 x 1 mL |
| Synonym | Dihydrotestosterone Undecanoate, (5alpha,17beta)-17-[(1-Oxoundecyl)oxy]androstan-3-one, Androstanolone Undecanoate |
| InChI Formula | InChI=1S/C30H50O3/c1-4-5-6-7-8-9-10-11-12-28(32)33-27-16-15-25-24-14-13-22-21-23(31)17-19-29(22,2)26(24)18-20-30(25,27)3/h22,24-27H,4-21H2,1-3H3/t22-,24-,25-,26-,27-,29-,30-/m0/s1 |
| SMILES | C[C@@]12[C@@]3([C@]([C@]4([C@](C)(CC3)[C@@H](OC(CCCCCCCCCC)=O)CC4)[H])(CC[C@]1(CC(=O)CC2)[H])[H])[H] |
| IUPAC Name | [(5S,8R,9S,10S,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl] undecanoate |
| Molecular Weight (g/mol) | 458.72 |
| Formula Weight | 458.376 |
| Concentration or Composition (by Analyte or Components) | 5alpha-Dihydro Testosterone Undecanoate 1 mg/mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?