missing translation for 'onlineSavingsMsg'
Learn More

(5Alpha)-4,5-Epoxy-14-hydroxy-2-iodo-3-methoxy-17-(2-propen-1-yl)-morphinan-6-one Cyclic 1,2-Ethanediyl Acetal, TRC

Product Code. 30388606
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388606 50 mg 50mg
30379069 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388606 Supplier Toronto Research Chemicals Supplier No. TRCE58964050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5alpha)-4,5-Epoxy-14-hydroxy-2-iodo-3-methoxy-17-(2-propen-1-yl)-morphinan-6-one Cyclic 1,2-Ethanediyl Acetal
Analyte or Component Names (5alpha)-4,5-Epoxy-14-hydroxy-2-iodo-3-methoxy-17-(2-propen-1-yl)-morphinan-6-one Cyclic 1,2-Ethanediyl Acetal
Shipping Condition Room Temperature
Molecular Formula C22H26INO5
InChI Formula InChI=1S/C22H26INO5/c1-3-7-24-8-6-20-16-13-11-14(23)17(26-2)18(16)29-19(20)22(27-9-10-28-22)5-4-21(20,25)15(24)12-13/h3,11,15,19,25H,1,4-10,12H2,2H3/t15-,19?,20+,21-/m1/s1
Solution Type Neat
SMILES IC1=CC2=C([C@@]3(C4C5(OCCO5)CC6)[C@]6(O)[C@H](N(CC=C)CC3)C2)C(O4)=C1OC
IUPAC Name (4'R,4a'S,12b'S)-3'-allyl-10'-iodo-9'-methoxy-1',2',3',4',5',6'-hexahydro-4a'H,7a'H-spiro[[1,3]dioxolane-2,7'-[4,12]methanobenzofuro[3,2-e]isoquinolin]-4a'-ol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 511.35
Quantity 50 mg
Formula Weight 511.09
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.