missing translation for 'onlineSavingsMsg'
Learn More
Learn More
58K Golgi Protein Antibody (FTCD/357) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14274JF525
This item is not returnable.
View return policy
Description
58K Golgi Protein Monoclonal antibody specifically detects 58K Golgi Protein in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| 58K Golgi Protein | |
| Monoclonal | |
| Janelia Fluor 525 | |
| formimidoyltransferase cyclodeaminase, formimidoyltransferase-cyclodeaminase, formiminotransferase cyclodeaminase, Formiminotransferase-cyclodeaminase, human formiminotransferase cyclodeaminase, EC 4.3.1.410formiminotransferase-cyclodeaminase, LCHC1 | |
| 58K Golgi Protein Protein purified from rat liver. (Uniprot: O95954) | |
| 0.1 mL | |
| Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism, Phospho Specific, Signal Transduction | |
| 10841 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| FTCD/357 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction