missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
58K Golgi Protein Monoclonal antibody specifically detects 58K Golgi Protein in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | 58K Golgi Protein |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | FTCD/357 |
| Conjugate | HRP |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | formimidoyltransferase cyclodeaminase, formimidoyltransferase-cyclodeaminase, formiminotransferase cyclodeaminase, Formiminotransferase-cyclodeaminase, human formiminotransferase cyclodeaminase, EC 4.3.1.410formiminotransferase-cyclodeaminase, LCHC1 |
| Host Species | Mouse |
| Immunogen | 58K Golgi Protein Protein purified from rat liver. (Uniprot: O95954) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?