missing translation for 'onlineSavingsMsg'
Learn More
Learn More
53BP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5110511
This item is not returnable.
View return policy
Description
Immunogen sequence: RALKGHVEQK RLSNGKLVEE IEQMNNLFQQ KQRELVLAVS KVEELTRQLE MLKNGRIDSH HDNQSAVAEL DRLYKELQLR NKLNQEQNAK LQQQRECLNK RNSEVAVMDK RVNELRDRLW KKKAALQQKE N
Regulator that plays a central role in regulation of apoptosis and cell growth via its interactions. Regulates TP53 by enhancing the DNA binding and transactivation function of TP53 on the promoters of proapoptotic genes in vivo. Inhibits the ability of APPBP1 to conjugate NEDD8 to CUL1, and thereby decreases APPBP1 ability to induce apoptosis. Impedes cell cycle progression at G2/M. Its apoptosis-stimulating activity is inhibited by its interaction with DDX42.
Specifications
| 53BP2 | |
| Polyclonal | |
| Unconjugated | |
| TP53BP2 | |
| 53BP2; Apoptosis-stimulating of p53 protein 2; apoptosis-stimulating protein of p53, 2; ASPP2; BBP; Bcl2-binding protein; P PPP1R13A; P53BP2; PPP1R13A; Renal carcinoma antigen NY-REN-51; TP53BP2; transformation related protein 53 binding protein 2; Trp53bp2; tumor protein p53 binding protein 2; tumor protein p53 binding protein, 2; tumor suppressor p53-binding protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7159 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 2.4 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q13625 | |
| TP53BP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-330 of TP53BP2 (NP_0010268552) | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction