missing translation for 'onlineSavingsMsg'
Learn More
Learn More
53BP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | 53BP2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227765
|
Novus Biologicals
NBP3-37959-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232108
|
Novus Biologicals
NBP3-37959-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
53BP2 Polyclonal antibody specifically detects 53BP2 in Human samples. It is validated for ELISA,Western BlotSpecifications
| 53BP2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, DNA Repair, Tumor Suppressors | |
| PBS (pH 7.3), 50% glycerol | |
| 7159 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| apoptosis-stimulating protein of p53, 2, ASPP2P53BP2,53BP2apoptosis-stimulating of p53 protein 2, Bbp, Bcl2-binding protein, p53-binding protein 2, p53BP2, PPP1R13A, Renal carcinoma antigen NY-REN-51, tumor protein p53 binding protein, 2, tumor protein p53-binding protein, 2, Tumor suppressor p53-binding protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-330 of TP53BP2 (NP_001026855.2).,, Sequence:, RALKGHVEQKRLSNGKLVEEIEQMNNLFQQKQRELVLAVSKVEELTRQLEMLKNGRIDSHHDNQSAVAELDRLYKELQLRNKLNQEQNAKLQQQRECLNKRNSEVAVMDKRVNELRDRLWKKKAALQQKEN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title