missing translation for 'onlineSavingsMsg'
Learn More

53BP2 Antibody (1A10), Novus Biologicals™

Product Code. 18327697 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327697 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327697 Supplier Novus Biologicals Supplier No. H00007159M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

53BP2 Monoclonal antibody specifically detects 53BP2 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen 53BP2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1A10
Conjugate Unconjugated
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH58918
Gene Alias apoptosis-stimulating protein of p53, 2, ASPP2P53BP2,53BP2apoptosis-stimulating of p53 protein 2, Bbp, Bcl2-binding protein, p53-binding protein 2, p53BP2, PPP1R13A, Renal carcinoma antigen NY-REN-51, tumor protein p53 binding protein, 2, tumor protein p53-binding protein, 2, Tumor suppressor p53-binding protein 2
Host Species Mouse
Immunogen TP53BP2 (AAH58918, 511 a.a. ~ 610 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PYFGQTNQPPSDIKPDGSSQQLSTVVPSMGTKPKPAGQQPRVLLSPSIPSVGQDQTLSPGSKQESPPAAAVRPFTPQPSKDTLLPPFRKPQTVAASSIYS
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7159
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.