missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 53BP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16334365 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16334365 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16334365 Supplier Invitrogen™ Supplier No. PA595093

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human RT4 whole cell. IHC: Human Intestinal Cancer tissue, Rat Brain tissue, Mouse Brain tissue. ICC/IF: A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 53BP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene TP53BP1
Gene Accession No. P70399, Q12888
Gene Alias 53BP1; FLJ41424; m53BP1; MGC138366; murine p53-binding protein; p202; p53-binding protein 1; p53BP1; TDRD30; TP53; TP53-binding protein 1; Tp53bp1; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; Trp53bp1; tumor protein 53-binding protein, 1; tumor protein p53 binding protein 1; tumor protein p53-binding protein, 1; tumor suppressor p53-binding protein 1
Gene Symbols TP53BP1, Trp53bp1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human 53BP1 (1955-1972aa ERIGFKQHPKYKHDYVSH).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27223, 296099, 7158
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.