missing translation for 'onlineSavingsMsg'
Learn More

53BP1 Antibody (1285C), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18416483 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416483 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416483 Supplier Novus Biologicals Supplier No. NBP254753AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

53BP1 Monoclonal antibody specifically detects 53BP1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen 53BP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone 1285C
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Host Species Rabbit
Immunogen 53BP1 Antibody (1285C) was made to a synthetic peptide made to the C-terminal portion of human 53BP1 protein (between amino acids 1900-1972) [UniProt Q12888]
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7158
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.