missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysyl Oxidase Homolog 2/LOXL2 Antibody (2E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26165-50ul
This item is not returnable.
View return policy
Description
Lysyl Oxidase Homolog 2/LOXL2 Monoclonal antibody specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Specifications
| Lysyl Oxidase Homolog 2/LOXL2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 2E5 | |
| Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 | |
| A synthesized peptide derived from Human Lysyl Oxidase Homolog 2/LOXL2 [UniProt Q9Y4K0] | |
| 50 μL | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 4017 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction