missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Complement C4 Antibody (SR3600), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27782-50ul
This item is not returnable.
View return policy
Description
Complement C4 Monoclonal antibody specifically detects Complement C4 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Complement C4 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR3600 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2, C4, complement component 4A (Rodgers blood group), RG | |
| A synthesized peptide derived from human Complement C4 (Uniprot #: P0C0L4) | |
| 50 μL | |
| Immunology, Innate Immunity | |
| 720 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction