missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin Antibody (4C1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26526-50ul
This item is not returnable.
View return policy
Description
Actin Monoclonal antibody specifically detects Actin in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| Actin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 4C1 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3 | |
| A synthesized peptide derived from Human Actin [UniProt P68133] | |
| 50 μL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Stem Cell Markers | |
| 58 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction