missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human PGLYRP4/PGRP-I beta Protein

Product Code. 18758937
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18758937

Brand: R&D Systems™ 8389PG050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human PGLYRP4/PGRP-I beta Protein is derived from HEK293. The Recombinant Human PGLYRP4/PGRP-I beta Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAK72485
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose
Gene ID (Entrez) 57115
Molecular Weight (g/mol) 40kDa
Name PGLYRP4/PGRP-I beta
Preparation Method Reconstitute at 500μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 53-64kDa, reducing conditions
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human PGLYRP4/PGRP-I beta protein Asp18-His373 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Asp18
Without Additives Carrier Protein
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.