missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human LILRB3/CD85a/ILT5 Protein

Product Code. 18777949
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18777949

Brand: R&D Systems™ 9159T5050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human LILRB3/CD85a/ILT5 is coated at 2 μg/mL, Recombinant Human Angiopoietin-like 7 (Catalog # 914-AN ) binds with an ED50 = 50-300 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAB68668
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 11025
Molecular Weight (g/mol) M.W. (Observed): 60-69 kDa, reducing conditions; M.W. (theoretical): 47 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human LILRB3/CD85a/ILT5 protein Gly24-Glu443, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD85 antigen-like family member A, CD85a, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3, LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB
Common Name ILT5/CD85a/LILRB3
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human LILRB3/CD85a/ILT5 is coated at 2μg/mL, Recombinant Human Angiopoietin-like 7 binds with an ED50 = 50-300ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Expression System Human embryonic kidney cell, HEK293-derived Gly24-Glu443, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ILT5/CD85a/LILRB3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.