missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human EGF-L6 Protein

Product Code. 18796167
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18796167

Brand: R&D Systems™ 8638EG050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human EGF-L6 Protein is derived from HEK293. The Recombinant Human EGF-L6 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 25975
Molecular Weight (g/mol) Observed M.W.: 40-55kDa, reducing conditions, Theoretical M.W.: 40kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human EGF-L6 protein, (N-terminus) Human EGF-L6 (Asn22-Arg363) Accession &Num; Q8IUX8 HP GGGSGGGSGGGS HHHHHH (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias DKFZp564P2063, EGF repeat-containing protein 6, EGFL6, EGF-like protein 6, EGF-like-domain, multiple 6, epidermal growth factor-like protein 6, MAEG, MAM and EGF domain containing, MAM and EGF domains-containing gene protein, W80
Common Name EGF-L6
Gene Symbol EGFL6
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of NIH-3T3 mouse embryonic fibroblast cells. Osada, A. et al. (2005) Exp. Cell Res. 303:148. The ED50 for this effect is typically 1-5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human EGF-L6 (Asn22-Arg363) Accession # Q8IUX8, N-terminus; HP; GGGSGGGSGGGS; HHHHHH; C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.