missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human C1qR1/CD93 Fc Chimera Avi-tag Protein, CF
Click to view available options
:
50 μg
Description
Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human C1qR1/CD93 Fc Chimera Avi-tag binds to Human IGFBP-rp1/IGFBP-7 (K95R) Protein (Catalog # 1334-B7 ) with an ED50 of 0.250-2.50 μg/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 22918 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 120-130 kDa, under reducing conditions., MolecularWeight-theroretical: 87 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human C1qR1/CD93 protein Human C1qR (Ala24-Lys580) (N-terminus) Accession # Q9NPY3.3 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | AA4 Antigen, C1q R1, C1q receptor 1, C1q Rp, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Collectin Receptor, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Ly68, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4 |
| Conjugate | Biotin |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction