missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSP Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00080331-B01P
This item is not returnable.
View return policy
Description
CSP Polyclonal antibody specifically detects CSP in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| CSP | |
| Polyclonal | |
| PBS (pH 7.4) | |
| CSP, Cysteine string protein, cysteine string protein alpha, DKFZp434N1429, DKFZp761N1221, DnaJ (Hsp40) homolog, subfamily C, member 5, dnaJ homolog subfamily C member 5, DNAJC5A, FLJ00118, FLJ13070 | |
| DNAJC5 (NP_079495.1, 1 a.a. - 198 a.a.) full-length human protein. MADQRQRSLSTSGESLYHVLGLDKNATSDDIKKSYRKLALKYHPDKNPDNPEAADKFKEINNAHAILTDATKRNIYDKYGSLGLYVAEQFGEENVNTYFVLSSWWAKALFVFCGLLTCCYCCCCLCCCFNCCCGKCKPKAPEGEETEFYVSPEDLEAQLQSDEREATDTPIVIQPASATETTQLTADSHPSYHTDGFN | |
| 50 μg | |
| Neuroscience | |
| 80331 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| NP_079495.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction