missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat EEA1 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF8047
This item is not returnable.
View return policy
Description
EEA1 Polyclonal specifically detects EEA1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry.
Specifications
| EEA1 | |
| Polyclonal | |
| Biotin | |
| Q15075 | |
| EEA1 | |
| E. coli-derived recombinant human EEA1 Asn1249-Gln1356 Accession # Q15075 | |
| 50 μg | |
| 8411 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunocytochemistry | |
| LYOPH | |
| Immunocytochemistry 15 ug/mL | |
| early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MST105, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2 | |
| Sheep | |
| Affinity Purified | |
| Primary | |
| EEA1 antibodies are ideal for immunocytochemistry colocalization studies in endosomes. The unconjugated antibody detects human, mouse, and rat EEA1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction