Learn More
Novus Biologicals™ Claudin-18.2 Antibody (zolbetuximab) - Chimeric, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28796-50ug
 View more versions of this product
Product Code. 30046492
  • £162.00 / 50µg
Description
Claudin-18.2 Monoclonal antibody specifically detects Claudin-18.2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claudin-18.2
Monoclonal
Unconjugated
P56856
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
zolbetuximab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
claudin-18, SFTA5, SFTPJ, surfactant associated 5, surfactant associated protein J, surfactant, pulmonary associated protein J
CLDN18.2
50 μg
Cell Biology, Cytoskeleton Markers, Signal Transduction
51208
Human
Purified