missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyanide (CN-) @ 1000 mg/L in 0.1% NaOH, VHG™
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
Description
Cyanide (CN-) @ 1000 mg/L in 0.1% NaOH
Specifications
Specifications
| CAS | 57-12-5 |
| Concentration | 1000 mg/L |
| Solvent or Matrix | Dilute NaOH |
| Quantity | 50 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction