missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XTT Sodium Salt, TRC
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
Specifications
Specifications
| Chemical Name or Material | [4-methoxy-5-[3-(2-methoxy-4-nitro-5-sulfonatophenyl)-5-(phenylcarbamoyl)tetrazol-3-ium-2-yl]-2-nitrophenyl]sulfonyloxysodium |
| CAS | 111072-31-2 |
| Molecular Formula | C22 H16 N7 Na O13 S2 |
| Quantity | 50 mg |
| Synonym | XTT Sodium Salt, Sodium 3'-[1-(Phenylaminocarbonyl)-3,4-tetrazolium]bis(4-methoxy-6-nitro)benzenesulfonic Acid, 2,3-Bis(2-methoxy-4-nitro-5-sulfophenyl)-5-[(phenylamino)carbonyl]-2H-tetrazolium Sodium Salt |
| InChI Formula | InChI=1S/C22H17N7O13S2.Na/c1-41-17-8-15(28(31)32)19(43(35,36)37)10-13(17)26-24-21(22(30)23-12-6-4-3-5-7-12)25-27(26)14-11-20(44(38,39)40)16(29(33)34)9-18(14)42-2;/h3-11H,1-2H3,(H2-,23,30,35,36,37,38,39,40);/q;+1/p-1 |
| SMILES | COc1cc(c(cc1[n+]2nc(nn2c3cc(c(cc3OC)[N+](=O)[O-])S(=O)(=O)O[Na])C(=O)Nc4ccccc4)S(=O)(=O)[O-])[N+](=O)[O-] |
| IUPAC Name | [4-methoxy-5-[3-(2-methoxy-4-nitro-5-sulfonatophenyl)-5-(phenylcarbamoyl)tetrazol-3-ium-2-yl]-2-nitrophenyl]sulfonyloxysodium |
| Molecular Weight (g/mol) | 673.521 |
| Formula Weight | 673.015 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction