missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Windaus Ketone, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Windaus Ketone |
| CAS | 55812-80-1 |
| Molecular Formula | C19 H32 O |
| Quantity | 50 mg |
| Synonym | 4H-Inden-4-one','octahydro-7a-methyl-1-[(1R,2E,4R)-1,4,5-trimethyl-2-hexen-1-yl]-','(1R,3aR,7aR)- |
| InChI Formula | InChI=1S/C19H32O/c1-13(2)14(3)8-9-15(4)16-10-11-17-18(20)7-6-12-19(16,17)5/h8-9,13-17H,6-7,10-12H2,1-5H3/b9-8+/t14-,15+,16+,17-,19+/m0/s1 |
| SMILES | CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2C(=O)CCC[C@]12C |
| IUPAC Name | (1R,3aR,7aR)-1-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-one |
| Molecular Weight (g/mol) | 276.46 |
| Formula Weight | 276.2453 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction