missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valeric Acid-d9, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Valeric Acid-d9 |
| CAS | 115871-50-6 |
| Molecular Formula | C5 D9 H O2 |
| Quantity | 50 mg |
| Synonym | 1-Butanecarboxylic Acid-d9, NSC 406833-d9, Propylacetic Acid-d9, n-Pentanoic Acid-d9, n-Valeric Acid-d9, Pentanoic-d9 acid (9CI), Pentanoic acid-d9, Valeric-d9 acid, Pentanoic-D9 acid |
| InChI Formula | InChI=1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i1D3,2D2,3D2,4D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,5-nonadeuteriopentanoic acid |
| Molecular Weight (g/mol) | 111.19 |
| Formula Weight | 111.1246 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction