missing translation for 'onlineSavingsMsg'
Learn More

Trihexyphenidyl N-Oxide, Mikromol™

Product Code. 30051688
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051688 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051688 Supplier Mikromol Supplier No. MM0237.04
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Trihexyphenidyl N-Oxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trihexyphenidyl N-Oxide
CAS 161564-79-0
Analyte or Component Names Trihexyphenidyl N-Oxide
Shipping Condition Room Temperature
Molecular Formula C20 H31 N O2
Certifications/Compliance ISO 9001
Synonym Trihexyphenidyl N-Oxide
InChI Formula InChI=1S/C20H31NO2/c22-20(18-10-4-1-5-11-18,19-12-6-2-7-13-19)14-17-21(23)15-8-3-9-16-21/h1,4-5,10-11,19,22H,2-3,6-9,12-17H2
SMILES OC(CC[N+]1([O-])CCCCC1)(C2CCCCC2)c3ccccc3
IUPAC Name 1-cyclohexyl-3-(1-oxidopiperidin-1-ium-1-yl)-1-phenylpropan-1-ol
Molecular Weight (g/mol) 317.47
Quantity 50 mg
Formula Weight 317.2355 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.