missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylenetetramine-d4 Tetrahydrochoride, TRC
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
Specifications
Specifications
| Chemical Name or Material | Triethylenetetramine-d4 Tetrahydrochoride |
| Molecular Formula | C6H14D4N4 • 4 (HCl) |
| Quantity | 50 mg |
| Synonym | N1,N2-Bis(2-aminoethyl)-1,2-ethanediamine-d4 Tetrahydrochoride, 1,2-Bis(2-aminoethylamino)ethane-d4 Tetrahydrochoride, 1,4,7,10-Tetraazadecane-d4 Tetrahydrochoride, 1,8-Diamino-3,6-diazaoctane-d4 Tetrahydrochoride, 3,6-Diazaoctane-1,8-diamine-d4 Tetrahydrochoride, Ancamine TETA-d4 Tetrahydrochoride, EpoxySet 145-20020-d4 Tetrahydrochoride, N,N'-Bis(2-aminoethyl)-1,2-diaminoethane-d4 Tetrahydrochoride, N,N'-Bis(2-aminoethyl)-1,2-ethanediamine-d4 Tetrahydrochoride, N,N'-Bis(2-aminoethyl)ethylenediamine-d4 Tetrahydrochoride, NSC 443-d4 Tetrahydrochoride, RT 1AX-d4 Tetrahydrochoride, Ren HY 956-d4 Tetrahydrochoride, Rutapox VE 2896-d4 Tetrahydrochoride, TECZA-d4 Tetrahydrochoride, TETA-d4 Tetrahydrochoride, Trien-d4 Tetrahydrochoride, Trientine-d4 Tetrahydrochoride, VE 2896-d4 Tetrahydrochoride, Z1 Tetrahydrochoride |
| InChI Formula | InChI=1S/C6H18N4.4ClH/c7-1-3-9-5-6-10-4-2-8;;;;/h9-10H,1-8H2;4*1H/i5D2,6D2;;;; |
| SMILES | NCCNC([2H])([2H])C([2H])([2H])NCCN.Cl.Cl.Cl.Cl |
| Molecular Weight (g/mol) | 150.2643646 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction