missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TAK 960 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TAK 960 hydrochloride is a potent PLK1 inhibitor (IC50 values are 0.8, 16.9 and 50.2 nM for PLK1, PLK2 and PLK3 respectively). Exhibits >20-fold selectivity for PLK1 over FAK, MLCK and FES, and has minimal activity against a panel of 282 other kinases. Inhibits proliferation of a range of cancer cell lines in vitro and inhibits tumor growth of multiple human cancer cell xenografts in vivo. Orally bioavailable.
Specifications
Specifications
| CAS | 2108449-45-0 |
| Quantity | 50 mg |
| Product Type | TAK 960 hydrochloride |
| Purity | >98% |
| Molecular Weight (g/mol) | 598.06 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction