missing translation for 'onlineSavingsMsg'
Learn More

Pomalidomide, MedChemExpress

Product Code. 16440657
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
5 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5g
This item is not returnable. View return policy

Product Code. 16440657

Brand: Medchem Express HY10984

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pomalidomide, the third-generation immunomodulatory agent, acts as molecular glue. Pomalidomide interacts with the E3 ligase cereblon and induces degradation of essential Ikaros transcription factors.

Pomalidomide, the third-generation immunomodulatory agent, acts as molecular glue. Pomalidomide interacts with the E3 ligase cereblon and induces degradation of essential Ikaros transcription factors.

Specifications

Chemical Name or Material Pomalidomide
CAS 19171-19-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H11N3O4
Quantity 50 mg
Synonym CC-4047
Solubility Information DMSO : 50 mg/mL (182.99 mM; Need ultrasonic)
SMILES O=C1N(C(C2=C1C=CC=C2N)=O)C(C(N3)=O)CCC3=O
Molecular Weight (g/mol) 273.24
Formula Weight 273.24
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.