missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piribedil, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
Description
Piribedil is a potent and orally active dopamine D2 and dopamine D3 agonist. Piribedil is also a α2-adrenoceptors antagonist. Piribedil can inhibit MLL1 methyltransferase activity (EC50: 0.18 μM). Piribedil has the potential for the research of parkinson's disease, circulatory disorders, cancers.
Specifications
Specifications
| Chemical Name or Material | Piribedil |
| CAS | 3605-01-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H18N4O2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 33.33 mg/mL (111.72 mM; Need ultrasonic) |
| SMILES | C1(N2CCN(CC3=CC=C(OCO4)C4=C3)CC2)=NC=CC=N1 |
| Molecular Weight (g/mol) | 298.34 |
| Formula Weight | 298.34 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction