missing translation for 'onlineSavingsMsg'
Learn More

Phenglutarimide Hydrochloride, Mikromol™

Product Code. 30051149
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30051149

Brand: Mikromol MM3210.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenglutarimide Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenglutarimide Hydrochloride
CAS 1674-96-0
Analyte or Component Names Phenglutarimide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H24 N2 O2 . Cl H
Certifications/Compliance ISO 17025
Synonym 2,6-Piperidinedione, 3-[2-(diethylamino)ethyl]-3-phenyl-, hydrochloride (1:1), 2,6-Piperidinedione, 3-[2-(diethylamino)ethyl]-3-phenyl-, monohydrochloride (9CI), Glutarimide, 2-[2-(diethylamino)ethyl]-2-phenyl-, hydrochloride (7CI), Glutarimide, 2-[2-(diethylamino)ethyl]-2-phenyl-, monohydrochloride (8CI), Aturbal, Aturban, Aturban hydrochloride, Aturbane, Phenglutarimide hydrochloride
InChI Formula InChI=1S/C17H24N2O2.ClH/c1-3-19(4-2)13-12-17(14-8-6-5-7-9-14)11-10-15(20)18-16(17)21;/h5-9H,3-4,10-13H2,1-2H3,(H,18,20,21);1H
SMILES Cl.CCN(CC)CCC1(CCC(=O)NC1=O)c2ccccc2
IUPAC Name 3-[2-(diethylamino)ethyl]-3-phenylpiperidine-2,6-dione;hydrochloride
Molecular Weight (g/mol) 324.85
Quantity 50 mg
Formula Weight 324.1605 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.