missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxcarbazepine, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Oxcarbazepine is a sodium channel blocker. Oxcarbazepine significantly inhibits glioblastoma cell growth and induces apoptosis or G2/M arrest in glioblastoma cell lines. Anti-cancer and anticonvulsant effects.
Specifications
Specifications
| Chemical Name or Material | Oxcarbazepine |
| Physical Form | Solid |
| CAS | 28721-07-5 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H12N2O2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | GP 47680 |
| Solubility Information | DMSO : 50 mg/mL (198.20 mM; Need ultrasonic) |
| Vis mere |
Research purposes only
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion